missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, 50% (w/w), Solution, 48-52%, Spectrum™ Chemical
$167.32 - $7,735.03
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18613615
|
Spectrum Chemical Mfg Cor
S1310100MLPL |
100 mL | Poly Bottle |
N/A
|
|
||||
18613616
|
Spectrum Chemical Mfg Cor
S1310500MLPL |
500 mL | Poly Bottle |
N/A
|
|
||||
18613617
|
Spectrum Chemical Mfg Cor
S13101LTPL |
1 L | Poly Bottle |
N/A
|
|
||||
18613618
|
Spectrum Chemical Mfg Cor
S13104LTPL |
4 L | Poly Bottle |
N/A
|
|
||||
18613620
|
Spectrum Chemical Mfg Cor
S131020LTPL |
20 L | Poly Drum |
N/A
|
|
||||
18613621
|
Spectrum Chemical Mfg Cor
S1310200LTBL |
200 L | Drum |
N/A
|
|
||||
18613619
|
Spectrum Chemical Mfg Cor
S131025GL |
2.5 gal. | Poly Drum |
N/A
|
|
||||
Chemical Identifiers
1310-73-2 |
Specifications
1310-73-2,7732-18-5 | |
48 to 52% | |
To Pass Test |
100 mL | |
Reagent | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.