missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, 98.5%, for analysis, pellets, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206060025

 View more versions of this product

Catalog No. AC206060025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium hydroxide
1390.0°C
318.0°C
HNaO
MFCD00003548
2.13g/mL
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Analytical
Pellet
White
1310-73-2
98.5%
NaOH
01,1083; 05,616; 07,336; 08,460
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
2.5 kg
[OH-].[Na+]
39.997
CHEBI:32145
98.5%
Plastic Bottle

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only