missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified, 50% (w/v) ±1%, LabChem™
$75.77 - $1083.93
Chemical Identifiers
| CAS | 1310-73-2 |
|---|---|
| Molecular Formula | HNaO |
| Molecular Weight (g/mol) | 39.997 |
| InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| PubChem CID | 14798 |
| ChEBI | CHEBI:32145 |
| IUPAC Name | sodium;hydroxide |
| SMILES | [OH-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC241401
|
LabChem
LC241401 |
500 mL |
Each for $75.77
|
|
|||||
|
LC241402
|
LabChem
LC241402 |
1 L |
Each for $122.23
|
|
|||||
|
LC241404
|
LabChem
LC241404 |
4 L |
Each for $141.77
|
|
|||||
|
LC241406
|
LabChem
LC241406 |
10 L |
Each for $563.73
|
|
|||||
|
LC241405
|
LabChem
LC241405 |
20 L |
Each for $1,083.93
|
|
|||||
Chemical Identifiers
| 1310-73-2 | |
| 39.997 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
| HNaO | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
Specifications
| Colorless | |
| 64.03,35.97 | |
| NaOH | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| 500 mL | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| 50% ±1% (w/v) | |
| Passes Test | |
| Traceable to NIST | |
| Sodium Hydroxide |
| 1310-73-2,7732-18-5 | |
| HNaO | |
| UN1824 | |
| Soluble in water | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40 | |
| Certified | |
| Poly Bottle | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Danger
Recommended Storage : Room Temperature