missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical™
Supplier: Thermo Fisher Scientific S39212
Specifications
14 | |
White | |
1310-73-2 | |
HNaO | |
1823 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
BP/EP/FCC/JP/NF | |
Poly Pail | |
1mmHg at 739°C |
Sodium Hydroxide | |
1390°C | |
318°C | |
MFCD00003548 | |
2.13g/cm³ | |
12 kg | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
95.0 to 100.5% (NF) | |
Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF) | |
Meets Requirements (BP), Meets Requirements (BP/EP/NF), Meets Requirements (EP), Meets Requirements (NF) | |
Solid |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive