missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide (Pellets/NF/FCC/EP/BP/JP), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S392212

 View more versions of this product

Catalog No. S392212


Explore available promotions

Description

Description

Specifications

Specifications

14
White
1310-73-2
HNaO
1823
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
BP/EP/FCC/JP/NF
Poly Bottle
1mmHg at 739°C
Sodium Hydroxide
1390°C
318°C
MFCD00003548
2.13g/cm³
2.5 kg
[OH-].[Na+]
39.997
CHEBI:32145
97.0 to 100.5% (BP)
Pass Test (BP), Pass Test (BP/EP/FCC/JP/NF), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (NF)
Meets Requirements (BP), Meets Requirements (BP/EP/NF), Meets Requirements (EP), Meets Requirements (NF)
Solid

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8 : Corrosive

SDS Documents

Documents

Promotions

Promotions