missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, Low Chloride, Pellets, ACS, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S129512KG

 View more versions of this product

Catalog No. 18613646


Explore available promotions

Description

Description

Specifications

Specifications

1310-73-2
HNaO
12 kg
sodium hydroxide
97%
To Pass Test
1
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
40.00
ACS
Poly Pail

Chemical Identifiers

1310-73-2
40.00
sodium hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.