Learn More
Sodium hydroxide monohydrate, 99.996% (metals basis)
CAS: 12179-02-1 | H3NaO2 | 58.01 g/mol
$63.38 - $824.40
Chemical Identifiers
| CAS | 12179-02-1 |
|---|---|
| Molecular Formula | H3NaO2 |
| Molecular Weight (g/mol) | 58.01 |
| MDL Number | MFCD00149229 |
| InChI Key | JBJWASZNUJCEKT-UHFFFAOYSA-M |
| Synonym | sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate |
| PubChem CID | 23719242 |
| SMILES | O.[OH-].[Na+] |
Description
Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12179-02-1 | |
| 58.01 | |
| JBJWASZNUJCEKT-UHFFFAOYSA-M | |
| 23719242 |
| H3NaO2 | |
| MFCD00149229 | |
| sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate | |
| O.[OH-].[Na+] |
Specifications
| White | |
| 12179-02-1 | |
| 99.996% (metals basis) | |
| MFCD00149229 | |
| sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate | |
| 5 g | |
| O.[OH-].[Na+] | |
| 58.01 | |
| 58.01 (40.00 Anhydrous) | |
| Crystals |
| 1390°C | |
| 318.4°C | |
| H3NaO2 | |
| 2.13 g/cm3 at 20°C | |
| Soluble in water and methanol. | |
| JBJWASZNUJCEKT-UHFFFAOYSA-M | |
| sodium;hydroxide;hydrate | |
| 23719242 | |
| Odorless | |
| Sodium hydroxide monohydrate |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 215-185-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only