missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, pellets, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals A1603736

Catalog No. AAA1603736


Explore available promotions

Description

Description

Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of
Specifications

Specifications

14
1390°C
318°C
2.13
500 g
Odorless
Sodium hydroxide
1310-73-2
MFCD00003548
Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
40

Chemical Identifiers

1310-73-2
MFCD00003548
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 215-185-5

RTECSNumber : WB4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only