missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Sodium Hydroxide, Pellets, BiotechGrade, 95-100.5%, Spectrum™ Chemical
S1301, 1310-73-2, NaOH
$448.56 - $6719.26
Identifiants chimiques
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 40.00 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
IUPAC Name | sodium hydroxide |
SMILES | [OH-].[Na+] |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | |||||
18606568
|
Spectrum Chemical Mfg Cor
S1301500GM |
500 g | Poly Bottle |
chaque for $448.56
|
|
|||||
18606570
|
Spectrum Chemical Mfg Cor
S130125KG |
2.5 kg | Poly Bottle |
chaque for $1,208.59
|
|
|||||
18606569
|
Spectrum Chemical Mfg Cor
S1301500GMCS6 |
6 x 500 g | Poly Bottle |
caisse de 6 chaque for $2,100.53
|
|
|||||
18606571
|
Spectrum Chemical Mfg Cor
S130125KGCS4 |
4 x 2.5 kg | Poly Bottle |
caisse de 4 chaque for $4,168.03
|
|
|||||
18606572
|
Spectrum Chemical Mfg Cor
S130112KG |
12 kg | Poly Pail |
chaque for $2,468.62
|
|
|||||
18606573
|
Spectrum Chemical Mfg Cor
S130150KGBL |
50 kg | Fiber Drum |
chaque for $6,719.26
|
|
|||||
Description
Spectrum™ Chemical Sodium Hydroxide, Pellets, BiotechGrade or caustic soda, is an inorganic, highly basic compound that is water soluble, as well as soluble in ethanol and methanol. One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.Identifiants chimiques
1310-73-2 | |
40.00 | |
sodium hydroxide |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] |
Spécifications
White | |
1 | |
500 g | |
[OH-].[Na+] | |
40.00 | |
Biotech | |
Poly Bottle | |
Sodium Hydroxide |
1310-73-2 | |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium hydroxide | |
95 to 100.5% | |
Odorless | |
Pellet |