missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, Pellets (Max. 0.02% K), For Analysis, EMSURE™, MilliporeSigma™
Supplier: Technidata 1064691000
Specifications
<14 (100g/L, H2O, 20°C) | |
White | |
319°C to 322°C | |
2.13Â g/3 (20Â °C) | |
1090g/L | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
ACS | |
Solid |
Sodium Hydroxide | |
1310-73-2 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
1 kg | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
99 to 100% | |
Plastic Bottle |
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
Safety and Handling
EINECSNumber : 215-185-5