missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, Pellets, NF, 95-100.5%, Spectrum™ Chemical
SO170, 1310-73-2, NaOH
$952.65 - $3187.85
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 40.00 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
IUPAC Name | sodium hydroxide |
SMILES | [OH-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18606582
|
Spectrum Chemical Mfg Cor
SO17025KG |
2.5 kg | Poly Bottle |
Each for $952.65
|
|
||||
18606581
|
Spectrum Chemical Mfg Cor
SO170500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $1,504.24
|
|
||||
18606583
|
Spectrum Chemical Mfg Cor
SO17025KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $3,170.50
|
|
||||
18606584
|
Spectrum Chemical Mfg Cor
SO17012KG |
12 kg | Poly Pail |
Each for $1,748.09
|
|
||||
18606585
|
Spectrum Chemical Mfg Cor
SO17050KGBL |
50 kg | Fiber Drum |
Each for $3,187.85
|
|
||||
Description
Spectrum™ Chemical Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
1310-73-2 | |
40.00 | |
sodium hydroxide |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
HNaO | |
2.5 kg | |
sodium hydroxide | |
95 to 100.5% | |
Poly Bottle |
1 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] | |
40.00 | |
NF |