missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, Pellets, NF, FCC, EP, JP, bioCERTIFIED™, 50 kg, Spectrum Chemical
Supplier: Spectrum Chemical Mfg Cor S940550KGBL
Specifications
Sodium Hydroxide | |
HNaO | |
50 kg | |
sodium hydroxide | |
95 to 97% | |
Poly Drum |
1310-73-2 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] | |
40.00 | |
NF/FCC/EP/JP | |
Pellets |
Chemical Identifiers
1310-73-2 | |
40.00 | |
sodium hydroxide |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] |