missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, Pellets, Technical, Spectrum™ Chemical
$199.85 - $3658.32
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18613654
|
Spectrum Chemical Mfg Cor
S1238500GM |
500 g | Poly Bottle |
Each for $199.85
|
|
|||||
18613655
|
Spectrum Chemical Mfg Cor
S123825KG |
2.5 kg | Poly Pail |
Each for $507.35
|
|
|||||
18613656
|
Spectrum Chemical Mfg Cor
S123812KG |
12 kg | Poly Pail |
Each for $1,477.41
|
|
|||||
18613657
|
Spectrum Chemical Mfg Cor
S123850KGBL |
50 kg | Fiber Drum |
Each for $3,658.32
|
|
|||||
Chemical Identifiers
1310-73-2 |
Specifications
1310-73-2 | |
500 g | |
Poly Bottle |
1 | |
Technical | |
Sodium Hydroxide |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.