missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution (1N/Certified), Fisher Chemical™

Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific BPSS2661

 View more versions of this product

Catalog No. SS266-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

13.3
Colorless
100°C
0°C
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Certified
Poly Bottle
Liquid
Sodium Hydroxide Solution
1N
1310-73-2,7732-18-5
HNaO
0.995 to 1.005N
1 L
[OH-].[Na+]
39.997
CHEBI:32145
Pass Test
LOT 84L Potassium acid phthalate

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes.
NFPA
Health:3 Flammability:0 Instability:0

Signal Word: DANGER!

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Accessories
Promotions

Promotions