missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical™
Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate
Supplier: Thermo Fisher Scientific FLSS26620
Specifications
13.3 | |
Colorless | |
100°C | |
0°C | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Certified | |
PolyPac™ | |
Liquid |
Sodium Hydroxide Solution | |
1N | |
1310-73-2,7732-18-5 | |
HNaO | |
0.995 to 1.005N | |
20 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Pass Test | |
LOT 84L Potassium acid phthalate |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
DANGER!
Emergency Overview
Causes burns by all exposure routes.
NFPA
Health:3 Flammability:0 Instability:0
Signal Word: DANGER!
DOTInformation : DOT Class 8, : Corrosive