missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 40%, Fisher Chemical™

Supplier:  Fisher Chemical SS4114

 Afficher plus de versions de ce produit

Numéro de catalogue. SS411-4



Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

12.0
40% (w/w)
1310-73-2,7732-18-5
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40 to 42%
Poly Bottle
Liquid
Sodium Hydroxide Solution
141.7°C
5°C
MFCD00003548
4 L
[OH-].[Na+]
39.997
CHEBI:32145
Pass Test
3mmHg at 37°C

Identifiants chimiques

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

DANGER!

Emergency Overview
Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1

missing translation for 'dotInformation' : DOT Class 8, : Corrosive

FDS Documents

Documents

Accessoires
Promotions

Promotions