missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hypochlorite, 5% Available Chlorine, Solution, ACS, Spectrum™ Chemical

$364.53 - $7266.85

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613676
View Documents
Spectrum Chemical Mfg Cor
S1669500ML
500 mL Poly Bottle
Each for $364.53
 
18613677
View Documents
Spectrum Chemical Mfg Cor
S16694LT
4 L Poly Bottle
Each for $1,416.15
 
18613678
View Documents
Spectrum Chemical Mfg Cor
S1669200LTBL
200 L Drum
Each for $7,266.85
 
Description

Description

Specifications

Chemical Identifiers

7681-52-9
74.44
sodium hypochlorite
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
[Na+].[O-]Cl

Specifications

7681-52-9,7732-18-5
500 mL
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium hypochlorite
5%
1
ClNaO
[Na+].[O-]Cl
74.44
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.