missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hypochlorite Solution, 5% available Chlorine, Ricca Chemical
Supplier: Ricca Chemical Company 7495532
Specifications
Sodium Hypochlorite Solution | |
7732-18-5,7681-52-9,7732-18-5 | |
96.0,6.12 | |
1 L | |
5% available Chlorine |
Colorless to Green-Yellow | |
92.0,5.88 | |
Liquid | |
dihydrogen oxide, dihydrogen monoxide | |
Amber Poly Bottle |
Chemical Identifiers
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide |
Safety and Handling
ShelfLife : 6 months