missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide, ACS, 99.5% min, Thermo Scientific Chemicals

Used to prevent iodine deficiency. Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines, in baking ingredient conditioner. Also used for conversion of alkyl chlorides into alkyl iodides

$147.38 - $454.71

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4019622
View Documents
Thermo Scientific Chemicals
04019622
100 g
Each for $147.38
 
AA4019636
View Documents
Thermo Scientific Chemicals
04019636
500 g
Each for $454.71
 
Description

Description

Sodium iodide commonly is used to prevent iodine deficiency. Used as analytical reagents, in the manufacture of disinfectants, antiseptics, deodorants and medicines. They are used in baking ingredient conditioner and as animal feed. It is used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
  • Commonly used to prevent iodine deficiency Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines
  • Used in baking ingredient conditioner and as animal feed
  • Used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements
Specifications

Chemical Identifiers

7681-82-5
149.894
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
CHEBI:33167
[Na+].[I-]

Specifications

1300°C to 1304°C
7681-82-5
Crystalline Powder
99.5% min.
MFCD00003532
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
Odorless
Hygroscopic
3.67 g/cm3 at 20°C
651°C
White
100 g
INa
14,8631
Soluble in water, alcohol, glycerol and acetone. Insoluble in ether.
[Na+].[I-]
149.894
CHEBI:33167
ACS
1.7745
White Powder
Sodium iodide
Videos
Safety and Handling

Safety and Handling

P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a: Avoid breathing dust/fume/gas/mist/vapours/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Take off contaminated clothing and wash before reuse. Call a POISON CENTER or doctor/physician if you feel unwell. Specific treatment (see label). Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-679-3

RTECSNumber : WB6475000

TSCA : Yes

storageNote1 : Hygroscopic and incompatible with strong acids,oxidizing agents. light, air and moisture sensitive.

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only