missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium iodide dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 202891000

 View more versions of this product

Catalog No. AC202891000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium iodide dihydrate
For analysis
White
Adhering Fine Crystals
99% min. (Argentometry)
NaI·2H2O
01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 14,89
sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
VEEIYHUOLCUPSA-UHFFFAOYSA-M
sodium;iodide;dihydrate
23685997
≥99%
Glass bottle
2.4400g/mL
752°C
13517-06-1
6 to 9 (5% aq. soln.)
100 g
INa·2H2O
MFCD00149211
10, 8469
Solubility in water: soluble. Other solubilities: soluble in alcohol,glycerol and acetone,very soluble in nh3
O.O.[Na+].[I-]
185.93
185.93
Analytical
2.44

Chemical Identifiers

13517-06-1
185.93
VEEIYHUOLCUPSA-UHFFFAOYSA-M
23685997
O.O.[Na+].[I-]
INa·2H2O
MFCD00149211
sodium iodide dihydrate, acmc-20ajo4, i.na.2h2o, sodium dihydrate iodide, sodium iodide-water 1/1/2, sodium iodide nai ,dihydrate 9ci
sodium;iodide;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only