missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium isopropoxide, Thermo Scientific Chemicals

$86.12 - $751.93

Chemical Identifiers

CAS 683-60-3
Molecular Formula C3H7NaO
Molecular Weight (g/mol) 82.078
MDL Number MFCD00061566
InChI Key WBQTXTBONIWRGK-UHFFFAOYSA-N
Synonym sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
PubChem CID 10920443
IUPAC Name sodium;propan-2-olate
SMILES CC(C)[O-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8944606
View Documents Promotion Details
Thermo Scientific Chemicals
08944606
5 g
Each for $86.12
 
AA8944614
View Documents Promotion Details
Thermo Scientific Chemicals
08944614
25 g
Each for $284.12
 
AA8944622
View Documents Promotion Details
Thermo Scientific Chemicals
08944622
100 g
Each for $751.93
 
Description

Description

Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

683-60-3
82.078
WBQTXTBONIWRGK-UHFFFAOYSA-N
10920443
CC(C)[O-].[Na+]
C3H7NaO
MFCD00061566
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
sodium;propan-2-olate

Specifications

683-60-3
C3H7NaO
5 g
sodium propan-2-olate, sodium isopropoxide, 2-propanol, sodium salt, isopropoxysodium, iprona, naoipr, 2-propanol, sodium salt 1:1, i-prona, nao-ipr, sodium isopropanolate
WBQTXTBONIWRGK-UHFFFAOYSA-N
sodium;propan-2-olate
10920443
Crystalline
70°C to 75°C
MFCD00061566
Air and moisture sensitive
Soluble in tetrahydrofuran.
CC(C)[O-].[Na+]
82.078
82.08
Sodium isopropoxide
Safety and Handling

Safety and Handling

P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314-H335-X

DOTInformation : Hazard Class: 4.2; Packaging Group: II

EINECSNumber : 211-673-7

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only