missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Laurate 97.0+%, TCI America™
$106.02 - $629.33
Chemical Identifiers
CAS | 629-25-4 |
---|---|
Molecular Formula | C12H23NaO2 |
Molecular Weight (g/mol) | 222.30 |
MDL Number | MFCD00041754 |
InChI Key | BTURAGWYSMTVOW-UHFFFAOYSA-M |
Synonym | sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026 |
PubChem CID | 2735067 |
IUPAC Name | sodium dodecanoate |
SMILES | [Na+].CCCCCCCCCCCC([O-])=O |
Chemical Identifiers
629-25-4 | |
222.30 | |
BTURAGWYSMTVOW-UHFFFAOYSA-M | |
2735067 | |
[Na+].CCCCCCCCCCCC([O-])=O |
C12H23NaO2 | |
MFCD00041754 | |
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026 | |
sodium dodecanoate |
Specifications
629-25-4 | |
C12H23NaO2 | |
25 g | |
sodium dodecanoate, sodium laurate, lauric acid sodium salt, dodecanoic acid, sodium salt, lauric acid, sodium salt, dodecanoic acid sodium salt, lauran sodny czech, caswell no. 778a, unii-k146mr5exo, epa pesticide chemical code 079026 | |
[Na+].CCCCCCCCCCCC([O-])=O | |
222.30 | |
222.30 | |
Crystalline Powder |
White | |
MFCD00041754 | |
3077 | |
BTURAGWYSMTVOW-UHFFFAOYSA-M | |
sodium dodecanoate | |
2735067 | |
≥97.0% (T) | |
Sodium Laurate |
Safety and Handling
EINECSNumber : (2)-0611&(7)-0973
RTECSNumber : OF0700000
TSCA : Yes