missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Linoleate 95.0+%, TCI America™

Supplier:  TCI America L00565G

 View more versions of this product

Catalog No. L00565G


Explore available promotions

Description

Description

Specifications

Specifications

Sodium Linoleate
192°C
C18H31NaO2
5 g
WYPBVHPKMJYUEO-UHFFFAOYSA-M
sodium;octadeca-9,12-dienoate
44134802
≥95.0% (T)
822-17-3
White-Yellow
MFCD00063202
Linoleic Acid Sodium Salt
CCCCCC=CCC=CCCCCCCCC(=O)[O-].[Na+]
302.434
302.43
Crystalline Powder

Chemical Identifiers

822-17-3
302.434
WYPBVHPKMJYUEO-UHFFFAOYSA-M
44134802
CCCCCC=CCC=CCCCCCCCC(=O)[O-].[Na+]
C18H31NaO2
MFCD00063202
Linoleic Acid Sodium Salt
sodium;octadeca-9,12-dienoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611&(7)-0973

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions