missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium meta-Periodate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S398100
Specifications
Sodium meta-Periodate | |
White | |
3.4 to 4.5 | |
100 g | |
INaO4 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
99.8 to 100.3% | |
Pass Test |
300°C | |
7790-28-5 | |
Solid | |
99.8 to 100.3 % | |
MFCD00003534 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium periodate | |
23667635 | |
213.89 | |
Certified ACS | |
Poly Bottle |
Chemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
sodium periodate |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 | |
[Na+].[O-][I](=O)(=O)=O |
Safety and Handling
DOTInformation : DOT Class 5.1, : Oxidizer