missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Metabisulfite (Powder/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S244500

 View more versions of this product

Catalog No. S244-500



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Metabisulfite
7681-57-4
4 to 6
500 g
Na2O5S2
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
[Na+].[Na+].[O-]S(=O)OS([O-])=O
190.09
190.11
Certified ACS
Glass Bottle
150°C
White
Powder/Solid
≥97 %
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
656671
≥97%
Pass Test
1.4g/cm³

Chemical Identifiers

7681-57-4
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
656671
[Na+].[Na+].[O-]S(=O)OS([O-])=O
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
disodium (sulfinatooxy)sulfinate
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Contact with acids liberates toxic gas. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions