Learn More
Sodium metaborate tetrahydrate, 98%, Thermo Scientific Chemicals
$63.55 - $219.60
Chemical Identifiers
CAS | 10555-76-7 |
---|---|
Molecular Formula | BH8NaO6 |
Molecular Weight (g/mol) | 137.86 |
MDL Number | MFCD00149205 |
InChI Key | JAKYJVJWXKRTSJ-UHFFFAOYSA-N |
Synonym | sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate |
PubChem CID | 23694267 |
IUPAC Name | sodium;oxido(oxo)borane;tetrahydrate |
SMILES | O.O.O.O.[Na+].[O-]B=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1180336
|
Thermo Scientific Chemicals
A1180336 |
500 g |
Each for $63.55
|
|
|||||
AAA118030E
|
Thermo Scientific Chemicals
A118030E |
2500 g |
Each for $219.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10555-76-7 | |
137.86 | |
JAKYJVJWXKRTSJ-UHFFFAOYSA-N | |
23694267 | |
O.O.O.O.[Na+].[O-]B=O |
BH8NaO6 | |
MFCD00149205 | |
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate | |
sodium;oxido(oxo)borane;tetrahydrate |
Specifications
∽53°C (decomposition) | |
500 g | |
MFCD00149205 | |
JAKYJVJWXKRTSJ-UHFFFAOYSA-N | |
sodium;oxido(oxo)borane;tetrahydrate | |
23694267 | |
98% | |
Sodium metaborate tetrahydrate |
10555-76-7 | |
BH8NaO6 | |
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate | |
O.O.O.O.[Na+].[O-]B=O | |
137.86 | |
137.86 (65.80 Anhydrous) | |
Odorless |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H319-H335-H361d
EINECSNumber : 231-891-6
RTECSNumber : ED4640000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only