missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium methanesulfonate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 442115000

 View more versions of this product

Catalog No. AC442115000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium methanesulfonate
500 g
CH3SO3Na
Solubility in water: soluble.
CS(=O)(=O)[O-].[Na+]
118.09
118.09
Authentic
2386-57-4
CH4NaO3S
sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
KKVTYAVXTDIPAP-UHFFFAOYSA-M
sodium;methanesulfonate
638112
99%

Chemical Identifiers

2386-57-4
118.09
sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
sodium;methanesulfonate
CH4NaO3S
KKVTYAVXTDIPAP-UHFFFAOYSA-M
638112
CS(=O)(=O)[O-].[Na+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 219-203-2

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only