missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Nitrite (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S347500

 View more versions of this product

Catalog No. S347-500

Explore available promotions

Description

Description

Specifications

Specifications

320°C
271°C
Yellow
Solid
≥97 %
MFCD00011118
LPXPTNMVRIOKMN-UHFFFAOYSA-M
sodium nitrite
23668193
69
Certified ACS
Poly Bottle
Sodium Nitrite
7632-00-0
8 to 9
500 g
NNaO2
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
[Na+].[O-]N=O
69.00
CHEBI:78870
≥97%
Pass Test
>320°C

Chemical Identifiers

7632-00-0
69.00
LPXPTNMVRIOKMN-UHFFFAOYSA-M
23668193
sodium nitrite
NNaO2
MFCD00011118
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
CHEBI:78870
[Na+].[O-]N=O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause central nervous system effects. May cause cyanosis. May cause methemoglobinemia. May cause skin, eye, and respiratory tract irritation. Very toxic to aquatic organisms. Use personal protective equipment. Ensure adequate ventilation. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:3
Flammability:0
Instability:2

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions