missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroprusside, Dihydrate, ACS Grade, 99.0 to 102.0%, LabChem™
Supplier: LabChem LC246909
Specifications
| Sodium Nitroprusside | |
| 13755-38-9 | |
| 100 | |
| 100 g | |
| Na2Fe(CN)5(NO)·2H2O | |
| UN1588 | |
| Soluble in water | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
| 297.95 | |
| 297.25 | |
| ACS | |
| Amber Glass | |
| Hydrogen cyanide; Carbon monoxide; Carbon dioxide |
| Dihydrate | |
| Brown | |
| Crystals | |
| C5H4FeN6Na2O3 | |
| MFCD00149192 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate | |
| 11953895 | |
| 99.0 to 102.0% | |
| Passes Test | |
| 1.72g/cm3 | |
| 1.72g/cm3 |
Chemical Identifiers
| 13755-38-9 | |
| 297.95 | |
| JFPDVRGGFBUORC-UHFFFAOYSA-N | |
| 11953895 | |
| O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
| C5H4FeN6Na2O3 | |
| MFCD00149192 | |
| nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
| disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature