missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium orthovanadate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 205330500

 View more versions of this product

Catalog No. AC205330500

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium orthovanadate
99%
White
50 g
Na3O4V
MFCD00003511
Solubility in water: soluble. Other solubilities: insoluble in ethanol
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
183.91
CHEBI:35607
99%
850.0°C to 866.0°C
13721-39-6
Powder
98.5% min. (as Na3VO4)
Na3VO4
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
IHIXIJGXTJIKRB-UHFFFAOYSA-N
trisodium;trioxido(oxo)vanadium
61671
183.91
Plastic bottle

Chemical Identifiers

13721-39-6
183.91
IHIXIJGXTJIKRB-UHFFFAOYSA-N
61671
trisodium;trioxido(oxo)vanadium
Na3O4V
MFCD00003511
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
CHEBI:35607
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 237-287-9

RTECSNumber : YW1120000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only