Learn More
Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1565622
Description
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.
Solubility
Soluble in water. Slightly soluble ethanol.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Sodium pentacyanonitrosylferrate(III) dihydrate | |
1.72 | |
C5H4FeN6Na2O3 | |
MFCD00149192 | |
UN1588 | |
14,8649 | |
Soluble in water. Slightly soluble ethanol. | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
297.95 | |
297.95 (261.92 Anhydrous) |
13755-38-9 | |
Odorless | |
Na2[Fe(CN)5NO]·2H2O | |
100 g | |
Hygroscopic | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate | |
11953895 | |
≥98% |
Chemical Identifiers
13755-38-9 | |
297.95 | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
11953895 | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
C5H4FeN6Na2O3 | |
MFCD00149192 | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate |
Safety and Handling
GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
EINECSNumber : 238-373-9
RTECSNumber : LJ8925000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only