missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium pentacyanonitrosylferrate(III) dihydrate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1565636

 View more versions of this product

Catalog No. AAA1565636


Explore available promotions

Description

Description

Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pentacyanonitrosylferrate(III) dihydrate, is used as a vasodilator drug to reduce blood pressure. It is used as an analytical reagent for the detection of methyl ketones and amines. Further, it is used as a reference compound for the calibration of Mossbauer spectrometer. It is also used for chromatographic detection of peptides containing cystine. It finds application as a color reagent in the development of thin layer chromatography in combination with acetaldehyde. It is also used in presence of buffers as reagent for ketone strips, which is utilized to test ketone in the urine of a diabetic.

Solubility
Soluble in water. Slightly soluble ethanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium pentacyanonitrosylferrate(III) dihydrate
1.72
C5H4FeN6Na2O3
MFCD00149192
UN1588
14,8649
Soluble in water. Slightly soluble ethanol.
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
297.95
297.95 (261.92 Anhydrous)
13755-38-9
Odorless
Na2[Fe(CN)5NO]·2H2O
500 g
Hygroscopic
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
JFPDVRGGFBUORC-UHFFFAOYSA-N
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
11953895
≥98%

Chemical Identifiers

13755-38-9
297.95
JFPDVRGGFBUORC-UHFFFAOYSA-N
11953895
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]
C5H4FeN6Na2O3
MFCD00149192
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P301+P310-P304+P340-P312-P314-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

EINECSNumber : 238-373-9

RTECSNumber : LJ8925000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only