Learn More
Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals
$67.58 - $206.36
Chemical Identifiers
CAS | 15630-89-4 |
---|---|
Molecular Formula | Na2CO3·1.5H2O2 |
MDL Number | MFCD00043204 |
Synonym | Hydrogen peroxide-Sodium carbonate adduct |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1604522
|
Thermo Scientific Chemicals
A1604522 |
100 g |
Each for $67.58
|
|
|||||
AAA1604536
|
Thermo Scientific Chemicals
A1604536 |
500 g |
Each for $74.14
|
|
|||||
AAA160450E
|
Thermo Scientific Chemicals
A160450E |
2500 g |
Each for $206.36
|
|
|||||
Description
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.
Chemical Identifiers
15630-89-4 | |
Na2CO3·1.5H2O2 | |
MFCD00043204 | |
Hydrogen peroxide-Sodium carbonate adduct |
Specifications
15630-89-4 | |
MFCD00043204 | |
UN1479 | |
Hydrogen peroxide-Sodium carbonate adduct | |
157.01 |
Na2CO3·1.5H2O2 | |
100 g | |
Moisture sensitive | |
Soluble in water. | |
Sodium percarbonate, 13 to 14% active oxygen |
Safety and Handling
GHS H Statement
H272-H318
May intensify fire; oxidizer.
Causes serious eye damage.
P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
H272-H302-H318
DOTInformation : Hazard Class: 5.1; Packaging Group: III
EINECSNumber : 239-707-6
RTECSNumber : FG0750000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only