Learn More
Sodium periodate, 99.8+%, ACS reagent
CAS: 7790-28-5 | INaO4 | 213.89 g/mol
$95.64 - $1187.40
Chemical Identifiers
| CAS | 7790-28-5 |
|---|---|
| Molecular Formula | INaO4 |
| Molecular Weight (g/mol) | 213.89 |
| MDL Number | MFCD00003534 |
| InChI Key | JQWHASGSAFIOCM-UHFFFAOYSA-M |
| Synonym | sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate |
| PubChem CID | 23667635 |
| ChEBI | CHEBI:75226 |
| SMILES | [Na+].[O-][I](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC419610050
|
Thermo Scientific Chemicals
419610050 |
5 g | Glass Bottle |
Each for $95.64
|
|
||||
|
AC419611000
|
Thermo Scientific Chemicals
419611000 |
100 g | Plastic bottle |
Each for $372.17
|
|
||||
|
AC419615000
|
Thermo Scientific Chemicals
419615000 |
500 g | Plastic bottle |
Each for $1,187.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 7790-28-5 | |
| 213.89 | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| 23667635 | |
| [Na+].[O-][I](=O)(=O)=O |
| INaO4 | |
| MFCD00003534 | |
| sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
| CHEBI:75226 |
Specifications
| 300.0°C | |
| White | |
| 5 g | |
| INaO4 | |
| MFCD00003534 | |
| 15,8772 | |
| Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents | |
| [Na+].[O-][I](=O)(=O)=O | |
| 213.89 | |
| CHEBI:75226 | |
| ≥99.8% | |
| Glass Bottle |
| 7790-28-5 | |
| Crystalline Powder | |
| 99.8 to 100.3% (on dried basis) (Iodometry) | |
| NaIO4 | |
| 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294 | |
| sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| sodium;periodate | |
| 23667635 | |
| 213.89 | |
| ACS Reagent | |
| Sodium periodate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
EINECSNumber : 232-197-6
RUO – Research Use Only