missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Peroxide (Certified ACS), Fisher Chemical

Supplier:  Thermo Fisher Scientific FLS363500

Catalog No. S363-500



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Peroxide
Yellow
Solid
≥93.0 %
MFCD00003497
PFUVRDFDKPNGAV-UHFFFAOYSA-N
disodium;peroxide
14803
≥93.0%
Pass Test
2.800g/cm³
460°C
1313-60-6,1310-73-2
500 g
Na2O2
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
[O-][O-].[Na+].[Na+]
77.978
77.98
Certified ACS
Tin Can

Chemical Identifiers

1313-60-6
77.978
PFUVRDFDKPNGAV-UHFFFAOYSA-N
14803
[O-][O-].[Na+].[Na+]
Na2O2
MFCD00003497
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
disodium;peroxide
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions