missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Persulfate (White Crystalline Solid/Certified), Fisher Chemical™
Supplier: Fisher Chemical O61141
Specifications
Sodium Persulfate | |
7775-27-1 | |
5 to 7 | |
1 kg | |
Na2O8S2 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] | |
238.092 | |
238.09 | |
Certified | |
Poly Bottle |
100°C | |
White | |
Powder/Solid | |
≥98.0 % | |
MFCD00003501 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
disodium;sulfonatooxy sulfate | |
62655 | |
≥98.0% | |
Pass Test | |
2.6g/cm³ |
Chemical Identifiers
7775-27-1 | |
238.092 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
62655 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
disodium;sulfonatooxy sulfate |
Safety and Handling
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause sensitization by inhalation and skin contact.
Signal Word: DANGER!
DOTInformation : DOT Class 5.1, : Oxidizer