missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Persulfate (White Crystalline Solid/Certified), Fisher Chemical™

Supplier:  Fisher Chemical O6114500

 View more versions of this product

Catalog No. O6114500



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Persulfate
7775-27-1
5 to 7
500 g
Na2O8S2
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
238.092
238.09
Certified
Poly Bottle
100°C
White
Powder/Solid
≥98.0 %
MFCD00003501
CHQMHPLRPQMAMX-UHFFFAOYSA-L
disodium;sulfonatooxy sulfate
62655
≥98.0%
Pass Test
2.6g/cm³

Chemical Identifiers

7775-27-1
238.092
CHQMHPLRPQMAMX-UHFFFAOYSA-L
62655
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]
Na2O8S2
MFCD00003501
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
disodium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause sensitization by inhalation and skin contact.

Signal Word: DANGER!

DOTInformation : DOT Class 5.1, : Oxidizer

SDS Documents

Documents

Promotions

Promotions