Learn More
Sodium phenoxide, 98%, Thermo Scientific Chemicals
$165.44 - $653.77
Chemical Identifiers
CAS | 139-02-6 |
---|---|
Molecular Formula | C6H5NaO |
Molecular Weight (g/mol) | 116.10 |
MDL Number | MFCD00013134 |
InChI Key | NESLWCLHZZISNB-UHFFFAOYSA-M |
Synonym | sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph |
PubChem CID | 4445035 |
ChEBI | CHEBI:52476 |
SMILES | [Na+].[O-]C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2500218
|
Thermo Scientific Chemicals
B2500218 |
50 g |
Each for $165.44
|
|
||||||
AAB2500230
|
Thermo Scientific Chemicals
B2500230 |
250 g |
Each for $653.77
|
|
||||||
Description
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.
Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
139-02-6 | |
116.10 | |
NESLWCLHZZISNB-UHFFFAOYSA-M | |
4445035 | |
[Na+].[O-]C1=CC=CC=C1 |
C6H5NaO | |
MFCD00013134 | |
sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph | |
CHEBI:52476 |
Specifications
139-02-6 | |
Phenol-like | |
C6H5ONa | |
50 g | |
Air Sensitive | |
Very soluble in water,soluble in alcoholSoluble in water,acetone and alcohol. | |
[Na+].[O-]C1=CC=CC=C1 | |
116.10 | |
CHEBI:52476 | |
98% |
>300°C | |
C6H5NaO | |
MFCD00013134 | |
UN2905 | |
sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph | |
NESLWCLHZZISNB-UHFFFAOYSA-M | |
sodium;phenoxide | |
4445035 | |
116.09 | |
Sodium phenoxide |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHENOLATES, SOLID
EINECSNumber : 205-347-3
RTECSNumber : SM8780000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only