missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phenylpyruvate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5676714
Description
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.
Solubility
Soluble in water
Notes
Store at 2-8°C. Avoin oxidizing agents.
Specifications
Sodium phenylpyruvate | |
>300°C | |
MFCD00002590 | |
4770978 | |
Soluble in water | |
C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+] | |
186.142 | |
186.14 |
114-76-1 | |
C9H7NaO3 | |
25 g | |
sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt | |
MQGYVGKMCRDEAF-UHFFFAOYSA-M | |
sodium;2-oxo-3-phenylpropanoate | |
23667645 | |
98% |
Chemical Identifiers
114-76-1 | |
186.142 | |
MQGYVGKMCRDEAF-UHFFFAOYSA-M | |
23667645 | |
C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+] |
C9H7NaO3 | |
MFCD00002590 | |
sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt | |
sodium;2-oxo-3-phenylpropanoate |
Safety and Handling
EINECSNumber : 204-053-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only