missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium phenylpyruvate monohydrate, 98%, Thermo Scientific Chemicals

$31.71 - $568.79

Chemical Identifiers

CAS 122049-54-1
Molecular Formula C9H7O3
Molecular Weight (g/mol) 163.15
MDL Number MFCD00149109
InChI Key BTNMPGBKDVTSJY-UHFFFAOYSA-M
Synonym sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
PubChem CID 23666336
IUPAC Name sodium;2-oxo-3-phenylpropanoate;hydrate
SMILES [O-]C(=O)C(=O)CC1=CC=CC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1402703
View Documents
Thermo Scientific Chemicals
A1402703
1 g
Each for $31.71
 
AAA1402706
View Documents
Thermo Scientific Chemicals
A1402706
5 g
Each for $90.79
 
AAA1402714
View Documents
Thermo Scientific Chemicals
A1402714
25 g
Each for $327.15
 
AAA1402718
View Documents
Thermo Scientific Chemicals
A1402718
50 g
Each for $568.79
 
Description

Description

Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

122049-54-1
163.15
BTNMPGBKDVTSJY-UHFFFAOYSA-M
23666336
[O-]C(=O)C(=O)CC1=CC=CC=C1
C9H7O3
MFCD00149109
sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
sodium;2-oxo-3-phenylpropanoate;hydrate

Specifications

122049-54-1
C9H7O3
MFCD00149109
4770978
Soluble in water.
[O-]C(=O)C(=O)CC1=CC=CC=C1
163.15
204.16 (186.14 Anhydrous)
Sodium phenylpyruvate monohydrate
>300°C
C6H5CH2COCO2Na·H2O
1 g
sodium phenylpyruvate monohydrate, sodium 2-oxo-3-phenylpropanoate hydrate, phenylpyruvic acid sodium salt monohydrate, phenylpyruvic acid, sodium salt, sodium hydrate ?-phenylpyruvate, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, acmc-2099lh, phenylpyruvic acid sodium hydrate, beta-phenylpyruvic acid, sodium salt
BTNMPGBKDVTSJY-UHFFFAOYSA-M
sodium;2-oxo-3-phenylpropanoate;hydrate
23666336
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-053-2

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only