missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™

Supplier:  Fisher BioReagents FLBP3321

 View more versions of this product

Catalog No. BP332-1



Explore available promotions

Description

Description

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)

This buffer component is commonly used in tissue culture and molecular biology applications.
Specifications

Specifications

Sodium Phosphate Dibasic Anhydrous
White
Powder/Solid
≥99.0 %
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
≥99.0%
0.2% max.
Decomposes information: > 240°C
7558-79-4
8.7 to 9.3
1 kg
HNa2O4P
15, 8789
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
24203
141.96
Pass Test
Poly Bottle

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
[Na+].[Na+].OP([O-])([O-])=O
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1

Signal Word: CAUTION!

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions