missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium phosphate dibasic dihydrate, 98+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 124992500

 View more versions of this product

Catalog No. AC124992500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium phosphate dibasic dihydrate
10028-24-7
98+%
Na2HPO4·2H2O
15,8789
(5% in water) Clear colorless
O.O.[Na+].[Na+].OP([O-])([O-])=O
177.99
177.99
Analytical
243.0°C to 245.0°C
250 g
H5Na2O6P
MFCD00149182
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
KDQPSPMLNJTZAL-UHFFFAOYSA-L
disodium;hydrogen phosphate;dihydrate
6451167
≥98%
Glass bottle

Chemical Identifiers

10028-24-7
177.99
KDQPSPMLNJTZAL-UHFFFAOYSA-L
6451167
O.O.[Na+].[Na+].OP([O-])([O-])=O
H5Na2O6P
MFCD00149182
disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
disodium;hydrogen phosphate;dihydrate
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only