missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis, Thermo Scientific Chemicals
$117.40 - $721.01
Chemical Identifiers
CAS | 10039-32-4 |
---|---|
Molecular Formula | HNa2O4P·12H2O |
Molecular Weight (g/mol) | 358.13 |
MDL Number | MFCD00149181 |
InChI Key | DGLRDKLJZLEJCY-UHFFFAOYSA-L |
Synonym | disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 |
PubChem CID | 61456 |
IUPAC Name | disodium;hydrogen phosphate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC271060010
|
Thermo Scientific Chemicals
271060010 |
1 kg | Plastic bottle |
Each for $117.40
|
|
||||
AC271060025
|
Thermo Scientific Chemicals
271060025 |
2.5 kg | Plastic bottle |
Each for $224.52
|
|
||||
AC271060100
|
Thermo Scientific Chemicals
271060100 |
10 kg | Plastic drum |
Each for $721.01
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10039-32-4 | |
358.13 | |
DGLRDKLJZLEJCY-UHFFFAOYSA-L | |
61456 | |
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] |
HNa2O4P·12H2O | |
MFCD00149181 | |
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 | |
disodium;hydrogen phosphate;dodecahydrate |
Specifications
35.0°C | |
10039-32-4 | |
Crystals | |
98.5% | |
Na2HPO4·12H2O | |
15, 8789 | |
Solubility in water: 218g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+] | |
358.13 | |
358.13 | |
Analytical | |
Sodium phosphate, dibasic dodecahydrate |
White | |
9 to 9.4 (5% aq. soln.) | |
1 kg | |
HNa2O4P·12H2O | |
MFCD00149181 | |
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12 | |
DGLRDKLJZLEJCY-UHFFFAOYSA-L | |
disodium;hydrogen phosphate;dodecahydrate | |
61456 | |
98.5% | |
Plastic bottle |
RUO – Research Use Only