missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Dibasic Heptahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical S373500
Specifications
| Sodium Phosphate Dibasic Heptahydrate | |
| White | |
| 9.0 to 9.3 | |
| 500 g | |
| H15Na2O11P | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| O.[Na+].[Na+].OP([O-])([O-])=O | |
| 268.06 | |
| 268.07 | |
| Certified ACS | |
| Poly Bottle |
| 48°C | |
| 7782-85-6 | |
| Solid | |
| 98 to 102 % | |
| MFCD00149180 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| disodium phosphoric acid heptahydrate | |
| 6096963 | |
| 98 to 102% | |
| Pass Test |
Chemical Identifiers
| 7782-85-6 | |
| 268.06 | |
| LVXHNCUCBXIIPE-UHFFFAOYSA-L | |
| 6096963 | |
| O.[Na+].[Na+].OP([O-])([O-])=O |
| H15Na2O11P | |
| MFCD00149180 | |
| sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
| disodium phosphoric acid heptahydrate |
Safety and Handling