missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium phosphate, monobasic, 99%, for biochemistry, anhydrous
CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
$129.20 - $272.37
Chemical Identifiers
| CAS | 7558-80-7 |
|---|---|
| Molecular Formula | H2NaO4P |
| Molecular Weight (g/mol) | 119.98 |
| MDL Number | MFCD00003527,MFCD00146206 |
| InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
| Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
| PubChem CID | 23672064 |
| ChEBI | CHEBI:37585 |
| IUPAC Name | sodium dihydrogen phosphate |
| SMILES | [Na+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC447982500
|
Thermo Scientific Chemicals
447982500 |
250 g | Plastic bottle |
Each for $129.20
|
|
||||
|
AC447980010
|
Thermo Scientific Chemicals
447980010 |
1 kg | Plastic bottle |
Each for $272.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| 23672064 | |
| sodium dihydrogen phosphate |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| CHEBI:37585 | |
| [Na+].OP(O)([O-])=O |
Specifications
| 7558-80-7 | |
| 99% | |
| NaH2PO4 | |
| 15,879 | |
| (1 M in water) Clear colorless | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| CHEBI:37585 | |
| 99% | |
| Plastic bottle |
| 250 g | |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium dihydrogen phosphate | |
| 23672064 | |
| 119.98 | |
| Biochemical | |
| Sodium phosphate, monobasic |
Safety and Handling
EINECSNumber : 231-449-2
RUO – Research Use Only