missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Monobasic, Anhydrous, Reagent Grade, LabChem™
Supplier: LabChem LC247751
Specifications
| Sodium Phosphate | |
| Monobasic, Anhydrous | |
| White | |
| Crystals or Granules | |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| Soluble in water | |
| [Na+].OP(O)([O-])=O | |
| 119.98 | |
| CHEBI:37585 | |
| Reagent | |
| Poly Bottle | |
| Phosphorus oxides |
| 203.9°C | |
| 7558-80-7 | |
| 100 | |
| 500 g | |
| NaH2PO4 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| sodium dihydrogen phosphate | |
| 23672064 | |
| 119.98 | |
| Passes Test | |
| 2.04g/cm3 | |
| 2.04g/cm3 |
Chemical Identifiers
| 7558-80-7 | |
| 119.98 | |
| AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
| 23672064 | |
| sodium dihydrogen phosphate |
| H2NaO4P | |
| MFCD00003527,MFCD00146206 | |
| monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
| CHEBI:37585 | |
| [Na+].OP(O)([O-])=O |
Safety and Handling
GHS H Statement
Product is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber : 231-449-2
Recommended Storage : Room Temperature