missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic monohydrate, 98%, extra pure

Sodium phosphate, monobasic monohydrate, 98%, H2NaO4P.H2O, CAS Number-10049-21-5 | CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol

$175.56 - $175.56

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC447740010
SDS View Documents
Thermo Scientific Chemicals
447740010
1 kg
Each for $175.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
H5NaO5P
MFCD00149208
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
O.[Na+].OP(O)(O)=O

Specifications

100.0°C
1 kg
H5NaO5P
MFCD00149208
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium;dihydrogen phosphate;hydrate
516949
98%
Plastic bottle
10049-21-5
98%
NaH2PO4·H2O
15,879
Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol
O.[Na+].OP(O)(O)=O
139.00
137.99
Extra Pure
Sodium phosphate, monobasic monohydrate
Videos
SDS
Documents

Documents

RUO – Research Use Only