Sodium Phosphate, Monobasic, Monohydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor S139550KGBL
Specifications
10049-21-5 | |
4.1 to 4.5 | |
98 to 102% | |
Fiber Drum |
1 | |
50 kg | |
ACS |
Chemical Identifiers
10049-21-5 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.