missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium polyphosphate, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 390930010

 View more versions of this product

Catalog No. AC390930010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium polyphosphate
White
Crystalline Powder
(NaO3P)n
MFCD00134118
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
[Na]OP(-*)(=O)O-*
65 to 70%
Plastic bottle
600.0°C
50813-16-6
1 kg
(NaPO3)n
15, 8773
Solubility in water: 1000g/l (20°C)
Sodium polyphosphate
Pure

Chemical Identifiers

50813-16-6
MFCD00134118
Sodium polyphosphate
(NaO3P)n
hexasodium oxido oxido phosphonatooxy phosphoryl oxy phosphoryl oxy phospho phosphonato oxy phosphinate
[Na]OP(-*)(=O)O-*
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 256-779-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only