Learn More
Sodium Potassium Tartrate Tetrahydrate (Crystalline/ ACS Grade), Fisher Chemical™
$468.67 - $5008.00
Chemical Identifiers
| CAS | 6381-59-5 |
|---|---|
| Molecular Formula | C4H12KNaO10 |
| Molecular Weight (g/mol) | 282.218 |
| MDL Number | MFCD00150989 |
| InChI Key | VZOPRCCTKLAGPN-UHFFFAOYSA-L |
| Synonym | potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j |
| PubChem CID | 2724148 |
| IUPAC Name | potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
| SMILES | C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
S387-500
|
Thermo Fisher Scientific
S387500 |
500 g | Poly Bottle |
Each for $468.67
Case of 6 Each for $2,812.02
|
|
||||
|
S387-3
|
Thermo Fisher Scientific
S3873 |
3 kg | Poly Bottle |
Each for $1,690.42
Case of 4 Each for $6,761.68
|
|
||||
|
S387-10
|
Thermo Fisher Scientific
S38710 |
10 kg | Poly Pail |
Each for $5,008.00
|
|
||||
Chemical Identifiers
| 6381-59-5 | |
| 282.218 | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| 2724148 | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] |
| C4H12KNaO10 | |
| MFCD00150989 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate |
Specifications
| 6381-59-5 | |
| 70°C | |
| 5.5 to 8.5 | |
| 99 to 102% | |
| C4H12KNaO10 | |
| 500 g | |
| 0.005% max. Insoluble Matter | |
| C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+] | |
| 282.218 | |
| 282.22 | |
| ACS | |
| Sodium Potassium Tartrate Tetrahydrate |
| >100°C | |
| White | |
| 437.22°C | |
| Poly Bottle | |
| MFCD00150989 | |
| potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j | |
| VZOPRCCTKLAGPN-UHFFFAOYSA-L | |
| potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate | |
| 2724148 | |
| 99 to 102% | |
| Powder or Solid |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0